man
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Reagents and Buffers
Electrophoresis Gel Casting Reagents
SeaPlaque™ GTG Agarose, Lonza™
Print
SeaPlaque™ GTG Agarose, Lonza™
744.00 € - 2210.00 €
See promotional offers below
Specifications
Chemical Name or Material
SeaPlaque GTG Agarose
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Quantity
Price
Quantity & Availability
Product Code
Brand
Quantity
Price
Quantity & Availability
11540737
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50111
25 g
744.00 €
25g
Estimated Shipment: 10-01-2025
Log in
to see stock.
Add to basket
11530737
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50110
125 g
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Specifications
Chemical Name or Material
SeaPlaque GTG Agarose
Videos
SDS
Documents
Documents
Product Certifications
Back to Top