Learn More
Idex Low-Pressure PEEK Tees
Combine, split, or intersect fluid streams throughout your system
Brand:  Idex P-712
 View more versions of this product
Product Code. 13060390
  • 55.15 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
Specifications
Specifications
Tees and Crosses
1000 psi
0.50 mm
0.020 in.
PEEK
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Idex Low-Pressure PEEK Tees > Tee; Natural; PEEK; Thru hole: 0.20 in.