missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPL Monoclonal Antibody (5D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518055
This item is not returnable.
View return policy
Description
This antibody does not recognize hepatic lipase and does not react with mouse samples. This target has a predicted molecular weight of 53 kDa.
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifications
| LPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P06858, P11151, P11153, P11602, Q06000 | |
| LPL | |
| Purified bovine milk lipoprotein lipase protein. | |
| 100 μL | |
| Primary | |
| Human, Rat, Bovine, Chicken, Guinea Pig | |
| Antibody | |
| IgG1 |
| ELISA, Inhibition Assays, Western Blot, Immunocytochemistry | |
| 5D2 | |
| Unconjugated | |
| LPL | |
| adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
| Mouse | |
| Protein G | |
| RUO | |
| 100135570, 24539, 280843, 396219, 4023 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction