Learn More
LW6, MedChemExpress
Product Code. 16408477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16408477
Brand: Medchem Express HY13671
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.
LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.
Specifications
Chemical Name or Material LW6
CAS 934593-90-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29NO5
Quantity 5 mg
Synonym HIF-1α inhibitor LW8
Solubility Information DMSO : 25 mg/mL (57.40 mM; Need ultrasonic) ∣DMF : 17.24 mg/mL (39.59 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(O)C(NC(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)=C1
Molecular Weight (g/mol) 435.51
Formula Weight 435.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death