missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies LY411575
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
- LY411575 is a cell-permeable γ-secretase inhibitor (IC50 = 0.14 nM) that blocks Notch activation in vitro at 500 μM (Curry et al.; Czirr et al.) via γ-Secretase inhibition
- Notch is a transmembrane receptor that plays a key role in cell fate decisions including cell proliferation, differentiation, and apoptosis
- Promotes neuronal differentiation of neural progenitor cells derived from mouse embryonic stem (ES) cells (Abranches et al.; Aranha et al.)
- Promotes goblet cell differentiation in mouse intestine and cultured colonic organoids (Okamoto et al.; Yui et al.)
- Induces hair cell differentiation from inner ear stem cells in vitro, and transdifferentiation of supporting cells into hair cells in vivo (Bramhall et al.; Mizutari et al.)
- Causes premature differentiation of Her4-positive progenitors into neurons in zebrafish (Dirian et al.)
- Induces apoptosis in primary and immortalized Karposi's sarcoma cells (Curry et al.)
Specifications
Specifications
| CAS | 209984-57-6 |
| Quantity | 5 mg |
| Molecular Formula | C26H23F2N3O4 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction