missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAGEA9 (Human) Recombinant Protein
Human MAGEA9 full-length ORF ( AAH02351.1, 1 a.a. - 315 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004108-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MSLEQRSPHCKPDEDLEAQGEDLGLMGAQEPTGEEEETTSSSDSKEEEVSAAGSSSPPQSPQGGASSSISVYYTLWSQFDEGSSSQEEEEPSSSVDPAQLEFMFQEALELKVAELVHFLLHKYRVKEPVTKAEMLESVIKNYKRYFPVIFGKASEFMQVIFGTDVKEVDPAGHSYILVTALGLSCDSMLGDGHSMPKAALLIIVLGVILTKDNCAPEEVIWEALSVMGVYVGKEHMFYGEPRKLLTQDWVQENYLEYRQVPGSDPAHYEFLWGSKAHAETSYEKVINYLVMLNAREPICYPSLYEEVLGEEQEGV
Specifications
AAH02351.1 | |
melanoma antigen family A, 9 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAGEA9 | |
GST |
4108 | |
Wheat germ expression system | |
25 μg | |
MAGE9, MGC8421 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |