Learn More
Applied Biosystems™ MagMAX™ Total RNA Elution Buffer
Product Code. 16316572 Shop All Applied Biosystems Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
For Research Use Only. All usage must comply with product instructions.
Product Code. 16316572
Brand: Applied Biosystems™ A41043
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
This elution buffer is included in the MagMAX-96 Total RNA Isolation Kit and MagMAX-96 Viral RNA Isolation Kit.
This elution buffer is included in the MagMAX-96 Total RNA Isolation Kit (Cat. No. AM1830) and MagMAX-96 Viral RNA Isolation Kit (AM1836). It is made available here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Product Line MagMAX
Quantity 10 mL
Shipping Condition Room Temperature