missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Malt Agar
BD Malt Agar for detection and isolation of yeasts and molds.
Brand: BD 224100
This item is not returnable.
View return policy
Description
- Malt agar is recommended for the cultivation, isolation, and maintenance of yeasts and molds.
- Testing conducted to identify the fungal impact on manufacturing processes
- Recognizes fungal isolates that may compromise human health
- Designed to contain the proper formulation essential for yeast and mold growth
Specifications
Dehydrated | |
Malt extract 30 g/L, Agar 15 g/L | |
Bottle |
Malt Agar | |
Dectection and Isolation of Yeasts and Molds | |
10 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Malt Agar > Malt Agar; Quantity: 10 kg
Spot an opportunity for improvement?Share a Content Correction