missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAOA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10539-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.Specifications
| MAOA | |
| Polyclonal | |
| Unconjugated | |
| MAOA | |
| MAO A, MAOA, monoamine oxidase A, Monoamine oxidase type A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4128 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21397 | |
| MAOA | |
| MAOA Fusion Protein Ag0821 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction