missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP2K5 (MEK5), GST Tag Recombinant Protein
Human MAP2K5 (MEK5), GST Tag Recombinant Protein
524.00 € - 7580.00 €
Specifications
Accession Number | AAH08838 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 76.4 kDa |
KinaseFamily | MAP Kinases |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17188745
|
Invitrogen™
A32895 |
100 μg |
2435.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15950303
|
Invitrogen™
A30977 |
10 μg |
524.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148795
|
Invitrogen™
A33380 |
1 mg |
7580.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2K5 (MEK5) serine/threonine kinase (mitogen-activated protein kinase kinase 5) or MEK5 is a dual specificity protein kinase that belongs to the MAP kinase kinase family which specifically interacts with and activates MAPK7/ERK5. MAP2K5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation. The rs3743354 polymorphism in the MAP2K5 promoter may affect the risk of developing colorectal cancer (CRC).- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
AAH08838 | |
76.4 kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Full Length, Recombinant, Active |
Kinase Assay | |
MAP Kinases | |
Human | |
GST Tag | |
Dry Ice | |
Liquid |