missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Marine Label Tape for BradyJet J4000 and J5000, BS 5609

Product Code. 17489202
Click to view available options
Width (Metric):
57.2 mm
101.6 mm
152.4 mm
203.2 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17489202

Brand: Brady™ 150762

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Don’t let a label on the final product sink your efficiency. With BS 5609 Marine Labels and the BradyJet J4000 or J5000 Printer, your labels stay stuck, even at sea.

  • Keep it colorful- this material runs through the BradyJet J4000 or J5000 Inkjet printer for high-resolution, full-color printing
  • An ink cartridge is required to print this material

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Continuous
Material Polyester (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Hazard Communication Identification, Marine Labeling
For Use With (Equipment) BradyJet J4000 and J5000 Color Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll/Roll
Quantity 1 Tape
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0088 in.
Thickness (Metric) 0.22 mm
Dimensions (LxW) 30.48 m x 152.4 mm
Length (English) 100 ft.
Length (Metric) 30.5 mm
Width (English) 6 in.
Width (Metric) 152.4 mm
Height (English) 100 ft.
Alternate Part Number J50C-6000-2585
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.