Learn More
Atago™ MASTER-BR Model Coolant Refractometer
Brand: Atago™ 2930
This item is not returnable.
View return policy
Specifications
Coolant Refractometer | |
MASTER-BR | |
Measuriing the concentration and freezing temperature of ethylene glycol and propylene glycol |
0 to 70% Ethylene Glycol, 0°C to -50°C, 0 to 70% Propylene Glycol, 0°C to -50°C | |
E.G/P.G | |
90 g |