Learn More
Masterflex™ Single-Channel Polycarbonate Housing Standard Pump Head
Click to view available options
Flow Rate:
0.002 to 8 L/min.
0.06 to 36 mL/min.
0.21 to 130 mL/min.
0.8 to 480 mL/min.
1.7 to 1000 mL/min.
2.8 to 1700 mL/min.
3.8 to 2300 mL/min.
4.8 to 2900 mL/min.
Rotor Material:
Cold-rolled Steel
Stainless Steel
Tubing Size:
13
14
15
16
18
19
24
35
36
73
82
Unit Size:
Each
Description
Use the pump heads for common fluid transfer applications requiring accuracy and excellent repeatability. MasterFlex™ Single-channel Polycarbonate Housing Standard Pump Heads are perfect for space-limited applications and easy to integrate into your OEM applications.
- High-precision economical pump head for precision tubing
- Standard heads are precision molded to accommodate a single size of tubing
- Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors
- CRS offers economy, while SS offers better chemical resistance and durability
- The 52 models with opaque black polyphenylene sulfide (PPS) housing and SS rotors offer the best overall chemical resistance
- Pump head is compact and simple in design with same side tubing entry/exit
- Standard heads are compact and easy to integrate into your OEM applications
Specifications
Specifications
| Dimensions (L x W x H) | 7.6 H x 7 W x 6.4 cm D |
| Flow Rate | 0.06 to 36 mL/min. |
| For Use With (Application) | For fluid transfer applications requiring high accuracy and repeatability |
| Includes | Tubing loading key, single-channel mounting hardware, and a 15 in. (38cm) length of Silicone pump tubing |
| No. of Channels | 1 |
| No. of Rollers | 3 |
| Occlusion Type | Fixed |
| Overall Depth (Metric) | 6.4 cm |
| Overall Height (Metric) | 7.6 cm |
| Overall Width (Metric) | 7 cm |
| Show More |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu