Learn More
Masterflex™ Tygon™ E-LFL Transparent Pump Tubing
Product Code. p-8001574
Click to view available options
Tubing Size:
13
14
15
17
18
26
35
70
73
82
88
89
Product Type:
I/P Pump Tubing
L/S Pump Tubing
Unit Size:
1 roll
Product Code. 12863383
Brand: Masterflex™ 0644013
Only null left
Add to basket
Add to basket
Use this high-performance pump tubing for challenging applications. Masterflex™ Transparent Tygon™ E-LFL Pump Tubing won’t age or oxidize, fits tightly to glass and metal and bends to small radius curves.
  • Exceeds USP class VI, 3A sanitary standards
  • FDA compliant
  • Nontoxic and non-hemolytic with excellent biocompatibility
  • Longest life of all Tygon formulations
  • Pump at continuous pressures up to 40psi
  • Low extractables and low gas permeability
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm
Material Tygon E-LFL
Overall Length (Metric) 762 cm
For Use With (Application) General laboratory use and provides longer life with peristaltic tubing pumps
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 1 Each
Tubing Size 13
Flow Rate 0.06 to 36 mL/min.
Hose Barb 1/16 in.
Height (Metric) 8.8 m
Gas Permeability Low
Color Transparent
Maximum Pressure 40 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type L/S Pump Tubing