Learn More
Mature Macrophage Marker Monoclonal Antibody (HIS36), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 12-0660-82
Description
Description: The HIS36 monoclonal antibody reacts with a molecule expressed on the surface of rat macrophages. The antigen recognized by HIS36 is an ED2-like antigen expressed by thioglycolate-elicited peritoneal cells and tissue macrophages. This antibody is used as a marker for rat tissue macrophages. Applications Reported: The HIS36 antibody has been reported for use in flow cytometric analysis. Applications Tested: The HIS36 antibody has been tested by flow cytometric analysis of rat spleen and peritoneal cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mature Macrophage Marker Monoclonal antibody specifically detects Mature Macrophage Marker in Rat samples. It is validated for Flow Cytometry.Specifications
Mature Macrophage Marker | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HIS36 | |
PE | |
ED2-like antigen; Macrophages; Mature Macrophages | |
Affinity chromatography | |
RUO | |
Rat | |
Antibody | |
IgG2a κ |
For Research Use Only.