Learn More
MBL™ Double Ended Borosilicate Glass Stirring Rod
Product Code. p-8002342
Click to view available options
Length (Metric):
200 mm
300 mm
Unit Size:
Pack of 10
Product Code. 12461627
Brand: MBL SRF386
Only null left
Add to basket
Add to basket
Minimize scratching and increase safety with these glass stirring rods. MBL™ Borosilicate Double Ended Glass Stirring Rods are ideal for a wide range of stirring applications.
  • Manufactured from borosilicate glass
  • With rounded button and paddle ends
  • Button end is useful for crushing clumped powder and solids
  • Paddle end helps to circulate sediment when stirring
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) General stirring applications
Includes Rounded button and paddle ends
Diameter (Metric) 6 mm
Length (Metric) 300 mm
Material Borosilicate Glass
Quantity 10/Pk.