missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid, 97+%

Product Code. 11414317
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11414317

Brand: Thermo Scientific Alfa Aesar B20391.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air Senstitive | CAS: 68-11-1 | C2H4O2S | 92.112 g/mol

Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

Solubility
Miscible with water, ethanol, ethers, ketones, esters, chlorinated hydrocarbons, benzene and aromatic hydrocarbons. Slightly miscible with chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-11-1
Molecular Formula C2H4O2S
Molecular Weight (g/mol) 92.112
MDL Number MFCD00004876
InChI Key CWERGRDVMFNCDR-UHFFFAOYSA-N
Synonym mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
PubChem CID 1133
ChEBI CHEBI:30065
IUPAC Name 2-sulfanylacetic acid
SMILES C(C(=O)O)S

Specifications

Melting Point -12°C
Density 1.295
Boiling Point 220°C
Flash Point 119°C (246°F)
Odor Stench
Linear Formula HSCH2CO2H
Refractive Index 1.5045
Quantity 2500 g
UN Number UN1940
Beilstein 506166
Sensitivity Air Sensitive
Merck Index 14,9336
Solubility Information Miscible with water,ethanol,ethers,ketones,esters,chlorinated hydrocarbons,benzene and aromatic hydrocarbons. Slightly miscible with chloroform.
Formula Weight 92.11
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Mercaptoacetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.