missing translation for 'onlineSavingsMsg'
Learn More

Dieldrin in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12935464
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 60-57-1
Packaging Ampoule(s)
Quantity 1 mL
Molecular Formula C12H8Cl6O
InChI Key DFBKLUNHFCTMDC-YNXGVQRSSA-N
SMILES C1C2C3C(C1C4C2O4)C5(C(=C(C3(C5(Cl)Cl)Cl)Cl)Cl)Cl
Molecular Weight (g/mol) 380.895
PubChem CID 101820673
Solvent or Matrix Methanol
Chemical Name or Material Dieldrin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.