missing translation for 'onlineSavingsMsg'
Learn More

Imazethapyr in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12937043
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 81335-77-5
Packaging Ampoule
Quantity 1 mL
Type Mass Spectrometry Standard
Molecular Formula C15H19N3O3
InChI Key XVOKUMIPKHGGTN-UHFFFAOYSA-N
SMILES CCC1=CN=C(C(=C1)C(=O)O)C2=NC(C(=O)N2)(C)C(C)C
IUPAC Name 5-ethyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol) 289.335
PubChem CID 54740
ChEBI CHEBI:82022
Grade EU Ph Ref: 1045200
Analyte or Component Names Imazethapyr
Solvent or Matrix Acetonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.