man
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Reagents and Buffers
Electrophoresis Gel Casting Reagents
MetaPhor™ Agarose
Print
MetaPhor™ Agarose
383.00 € - 3950.00 €
See promotional offers below
Specifications
Chemical Name or Material
Cambrex MetaPhor™ Agarose
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Quantity
Price
Quantity & Availability
Product Code
Brand
Quantity
Price
Quantity & Availability
11577795
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50181
25 g
383.00 €
25g
View alternative products
Save Up to
»
Estimated Shipment: 14-11-2024
Log in
to see stock.
Add to basket
11587795
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Lonza
50184
500 g
3950.00 €
500g
View alternative products
Save Up to
»
Estimated Shipment: 14-11-2024
Log in
to see stock.
Add to basket
Specifications
Chemical Name or Material
Cambrex MetaPhor™ Agarose
Videos
SDS
Documents
Documents
Product Certifications
Back to Top