missing translation for 'onlineSavingsMsg'
Learn More
Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals Product Code.: 10695412

Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals

Product Code. 10695412
250 mL, Glass bottle
Click to view available options
Quantity:
25 mL
250 mL
500 mL
1 L
4 L
25 L
Packaging:
Glass bottle
Metal Drum
Plastic drum
This item is not returnable. View return policy

Product Code. 10695412

Brand: Thermo Scientific Acros 168312500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-41-4, 150-76-5
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002651
InChI Key CERQOIWHTDAKMF-UHFFFAOYSA-N
Synonym methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methylShow MoreShow Less
PubChem CID 4093
ChEBI CHEBI:25219
IUPAC Name 2-methylprop-2-enoic acid
SMILES CC(=C)C(=O)O

Specifications

Melting Point 16.0°C
Density 1.0100g/mL
Boiling Point 163.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass bottle
Molecular Formula C4H6O2
Linear Formula H2C=C(CH3)CO2H
Refractive Index 1.4295 to 1.4315
Quantity 250 mL
Beilstein 02, 421
Merck Index 15, 6013
Specific Gravity 1.01
Stabilizer 250ppm (ca.) MEHQ
Formula Weight 86.09
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methacrylic acid, Stabilized
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464460 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168312500,168315000,168310010,168310040,168310251,168310250
Lot NumberA0466099 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number168312500,168315000,168310010,168310040,168310251,168310250
Lot NumberA0464460 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number168312500,168315000,168310010,168310040,168310251,168310250
Lot NumberA0457389 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number168312500,168315000,168310010,168310040,168310251,168310250
Lot NumberA0460304 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number168312500,168315000,168310010,168310040,168310251,168310250
compliance-icons compliance-icons
Product Identifier
  • Methacrylic acid, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.