Learn More
Methyl 2-acetamidoacrylate, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 35356-70-8 |
---|---|
Molecular Formula | C6H9NO3 |
Molecular Weight (g/mol) | 143.14 |
MDL Number | MFCD00013394 |
InChI Key | SMWNFFKPVLVOQQ-UHFFFAOYSA-N |
Synonym | methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate |
PubChem CID | 98644 |
IUPAC Name | methyl 2-acetamidoprop-2-enoate |
SMILES | COC(=O)C(=C)NC(C)=O |
Description
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
35356-70-8 | |
143.14 | |
SMWNFFKPVLVOQQ-UHFFFAOYSA-N | |
98644 | |
COC(=O)C(=C)NC(C)=O |
C6H9NO3 | |
MFCD00013394 | |
methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate | |
methyl 2-acetamidoprop-2-enoate |
Specifications
35356-70-8 | |
103°C to 104°C (8 mmHg) | |
MFCD00013394 | |
methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate | |
COC(=O)C(=C)NC(C)=O | |
143.14 | |
143.14 | |
Methyl 2-acetamidoacrylate |
49°C to 51°C | |
C6H9NO3 | |
1812208 | |
SMWNFFKPVLVOQQ-UHFFFAOYSA-N | |
methyl 2-acetamidoprop-2-enoate | |
98644 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only