missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals Product Code.: 15478238

Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 15478238
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15478238

Brand: Thermo Scientific Alfa Aesar H61675.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197507-22-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD09038282
InChI Key WYSPMXSNCAFCFV-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester
PubChem CID 22463192
IUPAC Name methyl 2-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)F

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluoro-4-hydroxybenzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81202671 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61675.MD,H61675.03,H61675.06
Lot NumberW17E062 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61675.MD,H61675.03,H61675.06
Lot NumberN28G017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61675.MD,H61675.03,H61675.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.