missing translation for 'onlineSavingsMsg'
Learn More
Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 11362399

Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 11362399
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11362399

Brand: Thermo Scientific Alfa Aesar H27318.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71486-53-8
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.627
MDL Number MFCD00012769
InChI Key NMAACQILAGCQPR-UHFFFAOYSA-N
Synonym methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
PubChem CID 2724028
IUPAC Name methyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point ∽175°C (decomposition)
Quantity 1 g
Beilstein 4273241
Formula Weight 193.63
Percent Purity 95%
Chemical Name or Material Methyl 4-oxopiperidine-3-carboxylate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.