Learn More
Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™
90.20 € - 576.00 €
Chemical Identifiers
CAS | 118465-49-9 |
---|---|
Molecular Formula | C8H7NO2S |
Molecular Weight (g/mol) | 181.209 |
MDL Number | MFCD07772830 |
InChI Key | YBIAQTPXONLBJG-UHFFFAOYSA-N |
Synonym | methyl 6h-thieno 2,3-b pyrrole-5-carboxylate, methyl thiopheno 3,2-d pyrrole-5-carboxylate, 5-methoxycarbonyl-6h-thieno 2,3-b pyrroleShow More |
PubChem CID | 7164601 |
IUPAC Name | methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate |
SMILES | COC(=O)C1=CC2=C(N1)SC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10331673
|
Thermo Scientific Maybridge
CC39322DA |
1 g | Amber Glass Bottle |
90.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239432
|
Thermo Scientific Maybridge
CC39322EA |
10 g | Amber Glass Bottle |
576.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
118465-49-9 | |
181.209 | |
YBIAQTPXONLBJG-UHFFFAOYSA-N | |
7164601 | |
COC(=O)C1=CC2=C(N1)SC=C2 |
C8H7NO2S | |
MFCD07772830 | |
methyl 6h-thieno 2,3-b pyrrole-5-carboxylate, methyl thiopheno 3,2-d pyrrole-5-carboxylate, 5-methoxycarbonyl-6h-thieno 2,3-b pyrroleShow More | |
methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate |
Specifications
118465-49-9 | |
100.0 | |
C8H7NO2S | |
methyl 6h-thieno 2,3-b pyrrole-5-carboxylate, methyl thiopheno 3,2-d pyrrole-5-carboxylate, 5-methoxycarbonyl-6h-thieno 2,3-b pyrrole | |
COC(=O)C1=CC2=C(N1)SC=C2 | |
181.209 | |
181.21 | |
Methyl 6H-thieno[2, 3-b]pyrrole-5-carboxylate |
96.5 | |
97% | |
MFCD07772830 | |
YBIAQTPXONLBJG-UHFFFAOYSA-N | |
methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate | |
7164601 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.