missing translation for 'onlineSavingsMsg'
Learn More
Methyl coumalate, 98%, Thermo Scientific Chemicals Product Code.: 11457006

Methyl coumalate, 98%, Thermo Scientific Chemicals

Product Code. 11457006
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11457006

Brand: Thermo Scientific Alfa Aesar B25330.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate. It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts. It has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates. It was also used in the preparation of 7-carboxyquinolizinium derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate. It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts. It has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates. It was also used in the preparation of 7-carboxyquinolizinium derivatives.

Solubility
Freely soluble in dichloromethane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-41-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00010120
InChI Key HHWWWZQYHPFCBY-UHFFFAOYSA-N
Synonym methyl coumalate, methyl 2-oxo-2h-pyran-5-carboxylate, coumalic acid, methyl ester, 5-carbomethoxy-2-pyrone, 2h-pyran-5-carboxylic acid, 2-oxo-, methyl ester, coumalic acid methyl ester, unii-1s7tmf0r8t, methyl 2-pyrone-5-carboxylate, methyl 2-oxopyran-5-carboxylate, 1s7tmf0r8t
PubChem CID 80113
IUPAC Name methyl 6-oxopyran-3-carboxylate
SMILES COC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 68°C to 72°C
Boiling Point 178°C to 180°C (60 mmHg)
Quantity 2 g
Merck Index 14,2557
Solubility Information Freely soluble in dichloromethane.
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material Methyl coumalate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243495 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog NumberB25330.04,B25330.09
Lot Number10164642 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25330.04,B25330.09
Lot Number10158457 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25330.04,B25330.09
Lot Number10195091 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25330.04,B25330.09
Lot Number10207023 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25330.04,B25330.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl coumalate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.