Learn More
Thermo Scientific Alfa Aesar METHYL CYCLOPROPANECARBOXYLATE, 98%,500G
Brand: Thermo Scientific Alfa Aesar B20853.36
316.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
METHYL CYCLOPROPANECARBOXYLATE, 98%,500G
Description

Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.