Learn More
Methyl Red, Solution 0.01%, Pure, Spirit Soluble, C.I 13020, Indicator Grade, Fisher Chemical
Product Code. 10020280
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
MDL Number 2425
Packaging Amber glass bottle
Quantity 500 mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Methyl red 0.01% solution, contains methylated spirit
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Acetone, Methyl alcohol,Sodium hydroxide