Learn More
Metro™ MetroMax i™ Top-Track Mobile Unit Kits
High-density, easy access storage
Brand:  Metro™ MXTTM18S
 View more versions of this product
Product Code. 15967221
  • 1150.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Includes: Posts, rigid casters, caster channels, bumpers and roller assemblies
This item is not returnable. View return policy
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
  • Order one per mobile unit
  • Shelves sold separately
Specifications
Specifications
Mobile Unit Kit
Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
45.7 cm
Metro™ MetroMax i™ and Q™ Top-Track™ system
18 in.