Learn More
Mettler Toledo™ Excellence RM Digital Benchtop Refractometers
Benchtop refractometers allow you to determine the refractive index, BRIX, HFCS, or user-defined concentrations of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence RM Digital Benchtop Refractometer has an in-built Peltier temperature control which provides both reliable temperature stability and rapid heating/cooling rates.
Brand:  Mettler Toledo™ 51337003
Product Code. 10468845
This item is not returnable. View return policy
Description
Description
  • Inert sapphire prism facilitates measurement of thin, highly volatile, dark, or viscous liquids
  • Removable top section provides ease of cleaning with simple tap water without the need for tools
  • Plug-and-play connection of accessories such as the printer, barcode scanner, LogStraight™ biometric fingerprint reader, and ErgoSens™ infrared sensor for hands-free operation
  • Assisted sample loading and clearing are possible through the addition of FillPal™ and DryPal™ units
  • Optional EQPac (Equipment Qualification Package) provides comprehensive, detailed qualification guidance (IQ/OQ/PQ) to those working in regulated environments
Expand Your Scope of Measurement
  • LiquiPhysics™ allows you to add parameters such as pH, color, and density/specific gravity to your refractive index readings
  • Simply connect a compatible SevenCompact™ pH or conductivity meter, colorimeter, or density cell to the RM module and define the method
  • Optional LabX™ LiquiPhysics™ Express software captures and manages all of these data points under a single platform
  • LabX™ also fully supports your CFR 21.11 requirements, provides user management, audit trail, flexible data export options, and fully customized reports
Reliable Standards
  • Mettler Toledo Certified Combined Density and Refractive Index Standards reduce risk, uncertainty, and error-prone sample transfer Standards come complete with certificates including an easy-to-scan barcode containing all information necessary for calibration
  • Barcodes save you valuable time, reduce data entry error, and improve traceability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0 to 100 Brix
Model RM40
20 sec.
5°C to 100°C
1.32 to 1.7
0.0001, 0.1% Brix
41°F to 212°F
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Excellence RM Digital Benchtop Refractometers > RM40; 4-place; 5.0° to 100.0°C