missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II Rat, PerCP-Cy5.5, Clone: M5/114.15.2, enQuireBio™

Rat Monoclonal Antibody

200.00 € - 545.00 €

Specifications

Antigen MHC Class II
Clone M5/114.15.2
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15962388
View Documents Promotion Details
EnquireBio
QAB70-PCP55-25ug
25 μg
200.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
15952388
View Documents Promotion Details
EnquireBio
QAB70-PCP55-100ug
100 μg
545.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

MHC Class II
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
14969
IgG2b κ
Protein A/G Purified
M5/114.15.2
Flow Cytometry
PerCP-Cyanine5.5
RUO
NP_034512
H2-Ab1 and H2-Eb1
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications