Learn More
Karl Hecht™ Micro Life™ Microscope Slides
Enable microscopic observation of long term cultures of microorganisms with this 'living slide.' Karl Hecht™ Micro Life™ Microscope Slides can be used with all light microscopes and stereoscopic magnifiers.
Brand: Karl Hecht™ 42426010
This item is not returnable.
View return policy
Description
- Observe long-term microorganism cultures.
- Can be used with all light microscopes and stereoscopic magnifiers.
- Correct plane location of coverglasses means that all microscopic contrast procedures are possible.
- Ideal for microscopic monitoring of activated sludge flocs in purification plants.
- Facilitates flow microscopy: e.g., plankton investigations or inorganic particles in solution can be reused after simple cleaning.
- Special cleaning and disinfecting solution Micro Life™ CLEANER No. 1403 suggested.
- Supplied with special pipette for filling vessels and flooding observation field.
- Sealing mode means that pump movements can be used for thorough and rapid mixing and cleaning procedures.
Specifications
Microscope Slide | |
Includes 1 Micro Life™ 2426 slide, 1 special pipette, operating instructions | |
Microscope |