Learn More
B Braun™ Micro Vascular Clip
Brand:  B Braun™ FD562R
Product Code. 10291460
  • 83.00 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
9 mm
Vascular Clip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
B Braun™ Micro Vascular Clip > Length: 9mm