Learn More
Applied Biosystems™ MicroAmp™ 3 x 32-Well Retainers
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 retainer
Unit Size:
Each
Description
- The Applied Biosystems MicroAmp 3 x 32-Well Retainer is specifically designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System.
- The retainer provides support and allows pressure to be distributed evenly over tubes and strips to ensure a proper seal and prevent the tubes and strips from being crushed or deformed.
- Its use is recommended at all times when working with MicroAmp reaction tubes and tube strips.
- Simply place the retainer on the sample block, then load the tubes or strips.
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Content And Storage | Store at room temperature. |
| Product Line | MicroAmp |
| Color | Black |
| Product Type | Retainer |
| For Use With (Equipment) | ProFlex 3 x 32-Well PCR System |
| For Use With (Application) | PCR |
| Plate Compatibility | 32-Well Plate |
| Quantity | 1 retainer |
| Shipping Condition | Room Temperature |