Learn More
Ansell™ Microchem™ 3000 Aprons with Sleeves
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Apron with Sleeves
- Rear velcro fastening
- Elasticated wrists
- Welded seams
- Multi-layer barrier fabric effective against numerous chemicals
- Highly visible bright yellow for improved worker safety
- Lightweight yet durable
- Anti-static according to EN 1149-5
Specifications
Specifications
Type | Protective Apron |
Size | Medium |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Closure Type | Hook and Loop (Rear) |
Cuff Style | Elastic |
No. of Pockets | None |
Certifications/Compliance | Type PB 3/4 EN 14605, Type PB(3)-B, PB(4)-B EN 14126, EN1149-1 |
Seam Type | Ultrasonically Welded |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ Microchem™ 3000 Aprons with Sleeves >
Spot an opportunity for improvement?Share a Content Correction