missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microsomal Glutathione S-transferase 1 Antibody (14H2-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32594
This item is not returnable.
View return policy
Description
Microsomal Glutathione S-transferase 1 Monoclonal antibody specifically detects Microsomal Glutathione S-transferase 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Microsomal Glutathione S-transferase 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 | |
| glutathione S-transferase 12, GST12EC 2.5.1.18, MGC14525, MGST, MGST-I, microsomal glutathione S-transferase 1, Microsomal GST-1, Microsomal GST-I | |
| Recombinant protein within Human Microsomal Glutathione S-transferase 1 aa 1-144/ 155. (Uniprot: P10620) | |
| 100 μL | |
| Cell Biology, Endocrinology | |
| 4257 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 14H2-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction