missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Microtube Insert

Grant Instruments Microtube insert used with MPS-1 High-speed shaker/vortex mixer.

56.18 €

Specifications

Material Polyethylene Foam EVA PE140 Base
Color Blue
For Use With (Equipment) MPS-1 High-Speed Shaker/Vortex Mixer
View More Specs

Products 4
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
15349625
View Documents Promotion Details
Grant Instruments™
P-02-05
24 x 0.5 mL PLUS 48 x 0.2 mL Tubes or 8 x 0.2 mL Strips
56.18 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
15359625
View Documents Promotion Details
Grant Instruments™
P-02-96
96 x 0.2 mL Tubes or 12 x 0.2 mL Strips or 96-well Semi/Unskirted PCR Plates
56.18 €
Each
In Stock
to see stock.
Add to basket
 
15369625
View Documents Promotion Details
Grant Instruments™
P-05-32
32 x 0.5 mL Tubes
56.18 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
15379625
View Documents Promotion Details
Grant Instruments™
P-2-24
24 x 1.5/2.0 mL Tubes
56.18 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Made from Polyethylene Foam EVA PE140
  • Color: Blue
  • For use with MPS-1 High-speed shaker/vortex mixer
  • Specifications

    Specifications

    Polyethylene Foam EVA PE140 Base
    Blue
    MPS-1 High-Speed Shaker/Vortex Mixer
    Videos
    SDS
    Documents

    Documents

    Product Certifications