Learn More
Invitrogen™ Mini Gel Tank Base
The Mini Gel Tank Base is a replacement part for the Mini Gel Tank. It holds the Gel Runner Tank.
Brand:  Invitrogen™ A25950
Product Code. 15344574
  • 33.30 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
This item is not returnable. View return policy
Description
Description
About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mini Gels
Mini
1 unit
Novex™ Mini Gels
Mini Gel Tank Base
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.