Learn More
MMAE-d8, MedChemExpress
MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Brand: Medchem Express HY-15162A
This item is not returnable.
View return policy
Description
MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.Specifications
MMAE-d8 | |
10 mg | |
C39H59D8N5O7 | |
Solid | |
White | |
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C | |
726.03 | |
Research |
2070009-72-0 | |
Research | |
Cancer-programmed cell death | |
Monomethyl auristatin E-d8 Deuterated labeled MMAE | |
DMSO : 100 mg/mL (137.74 mM; Need ultrasonic) | |
726.03 | |
98.43% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MMAE-d8, MedChemExpress