Learn More
Miele Professional™ Mobile unit
Product Code. 15460545 Shop All Miele Products
Click to view available options
:
Dimensions (D x W X H): 496 x 492 x 168mm
Product Code. 15460545
Brand: Miele Professional™ 5464630
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Module E 747 for optimum loading of centrifuge tubes, phials and test tubes, for example
  • 104 nozzles for centrifuge tubes, phials, test tubes, etc.
  • 104 nozzles 2.5 x 110 mm
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel, Plastic
Product Type Insert/Module for Test Tubes
Color Stainless Steel, Black
Holds 104 Test tubes
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 496 mm
Height (Metric) 168 mm
Width (Metric) 492 mm