Learn More
Miele Professional™ Mobile unit
Shop All Miele Products
Click to view available options
:
Dimensions (D x W X H): 790 x 640 x 565mm
Description
- The mobile unit can also be used without the upper level
- Magnetic strip for automatic mobile unit recognition
- Lower level equipped with a total of 115 nozzles with clips
Specifications
Specifications
| Material | Stainless Steel, Plastic |
| Product Type | Mobile Unit for Mesh Trays |
| Color | Stainless Steel, Black |
| Holds | 115 Narrow-necked glassware |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 790 mm |
| Height (Metric) | 565 mm |
| Width (Metric) | 640 mm |