Learn More
Memmert™ MobileALERT for UF55
Product Code. 17529445 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF55 Oven
Unit Size:
Each
Product Code. 17529445
Brand: Memmert™ UF55C3
Only null left
Add to basket
Add to basket
MobileALERT provides a notification by SMS with potential free contact error option.
  • MobileALERT
  • Notification by SMS in case of any error or alarm of the device
  • Requires option H6
Specifications
For Use With (Equipment) UF55 Oven
Type Alerting Device