missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBQCA Protein Quantitation Kit

Extremely sensitive assay for quantitating proteins in solution, capable of detection as low as 10ng of protein per mL

Brand:  Invitrogen™ C6667

Product Code. 10053032

This item is not returnable. View return policy

Description

Description

  • Similar in sensitivity to NanoOrange protein quantitation reagent (N-6666)
  • CBQCA is better suited for accurate quantitation of proteins in presence of lipids, membrane fractions or detergents, and for lipoproteins and small peptides

Protein Quantitation, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Fluorescence
Quantitating proteins in solution
1 Kit
Store in freezer (-5 to -30°C) and protect from light.
Fluorescent Protein Assay
Protein Quantitation Assay
Microplate Reader
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CBQCA Protein Quantitation Kit > CBQCA Protein Quantitation Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.