Learn More
Invitrogen™ CellLight™ Nucleus-RFP, BacMam 2.0
Provides easy way to label nuclei with red fluorescent protein in live cells
Brand: Invitrogen™ C10603
385.35 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses RFP fused to the SV40 nuclear localization sequence. You can observe nucleus-RFP behavior in live cells without the cellular toxicity associated with intercalators and label with multiple tracking or tracing dyes to image dynamic cellular processes.
Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.
BacMam Technology
CellLight Nucleus-RFP, BacMam 2.0, is a fusion construct of SV40 nuclear localization sequence and TagRFP, providing accurate and specific targeting to cellular nucleus-RFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.
- Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
- Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
- Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
- Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
- Flow Cytometer Laser Lines: 532
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cellular Imaging, Immunocytochemistry (ICC), Immunofluorescence (IF), Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing
Order Info
Shipping Condition: Wet Ice
Specifications
Red-Orange, Orange | |
555⁄584 | |
Confocal Microscope, Fluorescence Microscope | |
Fluorescence | |
Liquid | |
Wet Ice | |
Fluorescent Dye | |
Nucleus |
Store at 2°C to 6°C, protected from light. Do Not Freeze. | |
RFP (TagRFP) | |
1 vial | |
Visible | |
CellLight™ | |
Fluorescence Intensity | |
Nucleus Label |
For Research Use Only. Not for use in diagnostic procedures.