Learn More
Invitrogen™ FocalCheck™ Microspheres, 15 μm, fluorescent green ring stain/dark red throughout
Description
The sharp ring stains exhibited by the 15μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Specifications
Specifications
| Calibration Type | Confocal Microscope Calibration |
| Content And Storage | Store in refrigerator 2°C to 8°C and protect from light. |
| Color | Dark Red, Green |
| Product Type | Microsphere |
| Format | Suspension Beads |
| Product Line | FocalCheck |
| Quantity | 0.5 mL |
| Shipping Condition | Room Temperature |
| Diameter (Metric) | 15 μm |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.