Learn More
Invitrogen™ HCS LipidTOX™ Red Neutral Lipid Stain, for cellular imaging
Description
The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX Red neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX phospholipidosis detection reagents. HCS LipidTOX neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.
Cell Analysis, Cellular Imaging, High-Content Screening (HCS)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| For Use With (Equipment) | High Content Instrument |
| Quantity | 1 each |
| Detection Method | Fluorescence |
| Product Line | LipidTOX |
| Shipping Condition | Room Temperature |
| Label Type | Fluorescent Dye |
| Product Type | Lipid Stain |
| SubCellular Localization | Cell Membranes and Lipids, Lipids |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.