Learn More
Invitrogen™ SBFI, AM, cell permeant - Special Packaging
Sodium-sensitive molecule used to estimate Na+ gradients in isolated mitochondria, to measure intracellular NA+ levels, or to measure Na+ efflux in cells
Brand: Invitrogen™ S1264
963.68 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
- In combination with other fluorescent indicators used to correlate changes in intracellular Na+ with Ca2+ and Mg2+ concentrations, intracellular pH, and membrane potential
- Although selectivity of SBFI for Na+ is less than that of calcium indicators such as fura-2, it is sufficient for detection of physiological concentrations of Na+ in presence of other monovalent cations
- Spectral response of SBFI upon ion binding permit excitation ratio measurements, and this indicator can be used with same optical filters and equipment used for fura-
- Label (Ex/Em): SBFI (approximately 340,380/500nm)
- Lyophilized product may be dissolved in DMSO for use
- Product is typically loaded into cells by adding dissolved indicator to medium containing cells
- Selectivity Considerations and Cell Loading Strategies
- Dissociation constant (Kd) of SBFI for Na+ is 3.8mm in absence of K+, and 11.3mm in solutions with combined Na+ and K+ concentration of 135mm (which approximates physiological ionic strength)
- SBFI is approximately 18-fold more selective for Na+ than for K+
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cell-Based Ion Channel Assays, Drug Discovery and Development, Ion Channel Biology, Ionic Homeostasis and Signaling, Sodium and Potassium Detection, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature

Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Cell Viability and Proliferation | |
SBFI AM | |
20 x 50 μg |
Fluorescence | |
Fluorescence Microscope | |
Sodium Indicator | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
12Hours
23Minutes
27Seconds
31New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.