Learn More
Ibidi Mounting Medium with DAPI for Fluorescence Microscopy
Click to view available options
Quantity:
4
Unit Size:
Pack of 4
Description
- A mounting medium with DAPI that is optimized for fluorescence microscopy with Ibidi μ-Slides and μ-Dishes
- non-drying
- non-hardening
- volume stable
- 4x15 mL
Specifications
Specifications
| Quantity | 4 |
| Product Type | Mounting Medium |
| Volume (Metric) | 5 mL |