Learn More
Ibidi Mounting Medium with DAPI for Fluorescence Microscopy
Product Code. 17371329
Encompass_Preferred
Click to view available options
Quantity:
4
Unit Size:
Pack of 4
This item is not returnable. View return policy
Product Code. 17371329
Brand: Ibidi 500114
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Ibidi Mounting Medium with DAPI: for Fluorescence Microscopy, 4x15 mL is a mounting medium with DAPI that is optimized for fluorescence microscopy.
  • A mounting medium with DAPI that is optimized for fluorescence microscopy with Ibidi μ-Slides and μ-Dishes
  • non-drying
  • non-hardening
  • volume stable
  • 4x15 mL
Specifications
Quantity 4
Product Type Mounting Medium
Volume (Metric) 5 mL