Learn More
R&D Systems™ Mouse ASAM (NP_598494) VersaClone cDNA
R&D Systems™ Mouse ASAM (NP_598494) VersaClone cDNA is a type I transmembrane protein.
Brand: R&D Systems™ RDC2188
This item is not returnable.
View return policy
Description
The Mouse ASAM (NP_598494) VersaClone cDNA clone contains the complete ORF for ASAM, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ASAM insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mClmp | |
10 μg at 0.2 μg/μL | |
1135bp | |
79827 | |
Mouse |
NP_598494 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |