Learn More
Gibco™ Mouse FGF-7 (KGF) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 450-60-500
 View more versions of this product
Product Code. 30089081
  • 3145.00 € / Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P36363
Protein with no preservative
18.8
5 x 100 μg
-20°C
<1 EU/μg
FGF7
Unconjugated
CNDMSPEQTA TSVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNSYNIMEIR TVAVGIVAIK GVESEYYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHSGG EMFVALNQKG IPVKGKKTKK EQKTAHFLPM AIT
≥ 95% by SDS-PAGE gel and HPLC analyses.
Functional Assay
14178
Mouse FGF-7 (KGF)
E. coli
RUO
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Mouse
Recombinant
Lyophilized